Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286457.1 | NP_001273386.1 | 5 - 6 | 681 | 64 |
NM_001286458.1 | NP_001273387.1 | 6 - 7 | 856 | 64 | |
NM_001321858.1 | NP_001308787.1 | 5 - 6 | 607 | 64 | |
NM_003470.2 | NP_003461.2 | 5 - 6 | 812 | 64 | |
NR_135826.1 | 5 - 6 | 635 | 64 | ||
XM_017023652.1 | XP_016879141.1 | 5 - 6 | 1240 | 64 | |
XM_017023653.1 | XP_016879142.1 | 5 - 6 | 676 | 64 | |
GenBank mRNA | AK302481.1 | - | 6 - 7 | 782 | 64 |
AK302771.1 | - | 5 - 6 | 613 | 64 | |
AK302872.1 | - | 5 - 6 | 635 | 64 | |
AK302912.1 | - | 6 - 7 | 800 | 64 | |
AK316441.1 | - | 5 - 6 | 682 | 64 | |
AY376241.1 | - | 5 - 6 | 643 | 64 | |
Z72499.1 | - | 5 - 6 | 812 | 64 |