Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286457.1 | NP_001273386.1 | 7 - 8 | 922 | 101 |
NM_001286458.1 | NP_001273387.1 | 8 - 9 | 1097 | 101 | |
NM_001321858.1 | NP_001308787.1 | 7 - 8 | 848 | 101 | |
NM_003470.2 | NP_003461.2 | 7 - 8 | 1053 | 101 | |
NR_135826.1 | 7 - 8 | 876 | 101 | ||
XM_017023652.1 | XP_016879141.1 | 7 - 8 | 1481 | 101 | |
XM_017023653.1 | XP_016879142.1 | 7 - 8 | 917 | 101 | |
GenBank mRNA | AK302481.1 | - | 8 - 9 | 1023 | 101 |
AK302771.1 | - | 7 - 8 | 854 | 101 | |
AK302872.1 | - | 7 - 8 | 876 | 101 | |
AK302912.1 | - | 8 - 9 | 1041 | 101 | |
AK316441.1 | - | 7 - 8 | 923 | 101 | |
AY376241.1 | - | 7 - 8 | 884 | 101 | |
Z72499.1 | - | 7 - 8 | 1053 | 101 |