Product Details

Assay Reference Genome
Location

Chr.12:40249938 on build GRCh38
Cytoband
12q12
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
120892
Gene Symbol
LRRK2
Gene Name
leucine rich repeat kinase 2
Gene Aliases
AURA17, DARDARIN, PARK8, RIPK7, ROCO2
Location
Chr.12:40224895-40369285 on build GRCh38
Assay Gene Location
Overlaps Exon 8 - Intron 8
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LRRK2 NM_198578.3 NP_940980.3
XM_005268629.3 XP_005268686.1
XM_011537877.2 XP_011536179.1
XM_011537881.2 XP_011536183.1
XM_011537882.2 XP_011536184.1
XM_017018786.1 XP_016874275.1
XM_017018789.1 XP_016874278.1
AY792511.1 AAV63975.1
BC117180.1
BM285390.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1048258 Chr12:39970608 - 41209299 on Build GRCh38 Gain SLC2A13 PDZRN4 MUC19 LOC101930164 LRRK2 CNTN1
nsv1042967 Chr12:40235834 - 40316517 on Build GRCh38 Gain LRRK2

View Full Product Details