Assay Design
Gene Transcripts
- Gene Symbol
- ACCS
- Entrez Gene ID
-
84680
- Gene Name
- 1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
- Gene Aliases
-
ACS, PHACS
- Chromosome Location
-
Chr.11: 44066118 - 44084237 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK304074.1 |
- |
3 - 4 |
724 |
73 |
View Full Product Details