Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318360.1 | NP_001305289.1 | 14 - 15 | 2016 | 98 |
NM_003640.4 | NP_003631.2 | 14 - 15 | 2161 | 98 | |
XM_011519136.1 | XP_011517438.1 | 14 - 15 | 1953 | 98 | |
XM_011519137.1 | XP_011517439.1 | 14 - 15 | 1727 | 98 | |
XM_017015238.1 | XP_016870727.1 | 12 - 13 | 1732 | 98 | |
GenBank mRNA | AB210033.1 | - | 14 - 15 | 1923 | 98 |
AF044195.1 | - | 14 - 15 | 1940 | 98 | |
AF153419.2 | - | 14 - 15 | 1947 | 98 | |
AK001641.1 | - | 14 - 15 | 1945 | 98 | |
AK127237.1 | - | 6 - 7 | 1576 | 98 | |
AK289962.1 | - | 14 - 15 | 1953 | 98 | |
AK304742.1 | - | 14 - 15 | 1658 | 98 | |
BC033094.1 | - | 14 - 15 | 1723 | 98 | |
CR749385.1 | - | 7 - 8 | 988 | 98 |