Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318360.1 | NP_001305289.1 | 33 - 34 | 3950 | 78 |
NM_003640.4 | NP_003631.2 | 33 - 34 | 4095 | 78 | |
XM_011519136.1 | XP_011517438.1 | 33 - 34 | 3929 | 78 | |
XM_011519137.1 | XP_011517439.1 | 33 - 34 | 3703 | 78 | |
XM_017015238.1 | XP_016870727.1 | 31 - 32 | 3666 | 78 | |
GenBank mRNA | AB210033.1 | - | 33 - 34 | 3857 | 78 |
AF044195.1 | - | 33 - 34 | 3874 | 78 | |
AF153419.2 | - | 33 - 34 | 3881 | 78 | |
AK001641.1 | - | 33 - 34 | 3879 | 78 | |
AK022559.1 | - | 15 - 16 | 1564 | 78 | |
AK024223.1 | - | 13 - 14 | 1338 | 78 | |
AK123257.1 | - | 6 - 7 | 1651 | 78 | |
AK127237.1 | - | 25 - 26 | 3510 | 78 | |
AK225798.1 | - | 20 - 21 | 2179 | 78 | |
AK289962.1 | - | 33 - 34 | 3887 | 78 | |
AK304742.1 | - | 33 - 34 | 3592 | 78 | |
AL049945.1 | - | 7 - 8 | 692 | 78 | |
BC033094.1 | - | 33 - 34 | 3657 | 78 | |
CR749385.1 | - | 26 - 27 | 2922 | 78 |