Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318360.1 | NP_001305289.1 | 7 - 8 | 1026 | 104 |
NM_003640.4 | NP_003631.2 | 7 - 8 | 1171 | 104 | |
XM_011519136.1 | XP_011517438.1 | 7 - 8 | 963 | 104 | |
XM_011519137.1 | XP_011517439.1 | 7 - 8 | 737 | 104 | |
GenBank mRNA | AB210033.1 | - | 7 - 8 | 933 | 104 |
AF044195.1 | - | 7 - 8 | 950 | 104 | |
AF153419.2 | - | 7 - 8 | 957 | 104 | |
AK001641.1 | - | 7 - 8 | 955 | 104 | |
AK289962.1 | - | 7 - 8 | 963 | 104 | |
AK304742.1 | - | 7 - 8 | 668 | 104 | |
BC033094.1 | - | 7 - 8 | 733 | 104 |