Assay Design



Gene Transcripts

Gene Symbol
PLRG1
Entrez Gene ID
5356
Gene Name
pleiotropic regulator 1
Gene Aliases
Cwc1, PRL1, PRP46, PRPF46, TANGO4
Chromosome Location
Chr.4: 154534997 - 154550433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201564.1 NP_001188493.1 4 - 5 408 84
GenBank mRNA BC020786.1 - 4 - 5 372 84

View Full Product Details