Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000701.7 | NP_000692.2 | 3 - 4 | 517 | 135 |
NM_001160233.1 | NP_001153705.1 | 3 - 4 | 541 | 135 | |
NM_001160234.1 | NP_001153706.1 | 3 - 4 | 350 | 135 | |
XM_017001360.1 | XP_016856849.1 | 4 - 5 | 3736 | 135 | |
XM_017001361.1 | XP_016856850.1 | 4 - 5 | 3078 | 135 | |
GenBank mRNA | AK295095.1 | - | 3 - 4 | 540 | 135 |
AK296362.1 | - | 3 - 4 | 350 | 135 | |
AK296384.1 | - | 3 - 4 | 476 | 135 | |
AK308418.1 | - | 3 - 4 | 282 | 135 | |
AK314777.1 | - | 3 - 4 | 362 | 135 | |
BC003077.2 | - | 3 - 4 | 430 | 135 | |
BC050359.1 | - | 3 - 4 | 428 | 135 | |
BF795089.1 | - | - | 477 | 135 | |
DC335233.1 | - | 3 - 4 | 517 | 135 | |
U16798.1 | - | 2 - 3 | 178 | 135 |