Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000701.7 | NP_000692.2 | 7 - 8 | 1097 | 75 |
NM_001160233.1 | NP_001153705.1 | 7 - 8 | 1121 | 75 | |
NM_001160234.1 | NP_001153706.1 | 7 - 8 | 930 | 75 | |
XM_017001360.1 | XP_016856849.1 | 8 - 9 | 4316 | 75 | |
XM_017001361.1 | XP_016856850.1 | 8 - 9 | 3658 | 75 | |
GenBank mRNA | AK295095.1 | - | 7 - 8 | 1120 | 75 |
AK296362.1 | - | 7 - 8 | 930 | 75 | |
AK296384.1 | - | 7 - 8 | 1056 | 75 | |
AK308418.1 | - | 7 - 8 | 862 | 75 | |
AK314777.1 | - | 7 - 8 | 942 | 75 | |
BC003077.2 | - | 7 - 8 | 1010 | 75 | |
BC050359.1 | - | 7 - 8 | 1008 | 75 | |
J03007.1 | - | 2 - 3 | 167 | 75 | |
U16798.1 | - | 6 - 7 | 758 | 75 |