Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002472.2 | NP_002463.2 | 16 - 17 | 1988 | 106 |
GenBank mRNA | AK303395.1 | - | 16 - 17 | 1988 | 106 |
M36769.1 | - | 15 - 16 | 1966 | 106 | |
X51592.1 | - | 2 - 3 | 390 | 106 | |
Z38133.1 | - | 15 - 16 | 1966 | 106 |