Assay Design



Gene Transcripts

Gene Symbol
TEAD1
Entrez Gene ID
7003
Gene Name
TEA domain transcription factor 1
Gene Aliases
AA, NTEF-1, REF1, TCF-13, TCF13, TEAD-1, TEF-1
Chromosome Location
Chr.11: 12674422 - 12944737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021961.5 NP_068780.2 6 - 7 1085 65
GenBank mRNA AB209858.1 - 1 - 2 400 65
AL833289.1 - 7 - 8 694 65
BC115398.1 - 6 - 7 590 65

View Full Product Details