Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018420.2 | NP_060890.2 | 2 - 3 | 426 | 71 |
XM_005271004.3 | XP_005271061.1 | 2 - 3 | 387 | 71 | |
GenBank mRNA | AY145501.1 | - | 2 - 3 | 296 | 71 |
AY358258.1 | - | 2 - 3 | 310 | 71 | |
BC026358.1 | - | 2 - 3 | 425 | 71 | |
KU178698.1 | - | 2 - 3 | 260 | 71 | |
KU178699.1 | - | 2 - 3 | 260 | 71 | |
KU178700.1 | - | 2 - 3 | 260 | 71 | |
KU178702.1 | - | 2 - 3 | 260 | 71 |