Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000701.7 | NP_000692.2 | 15 - 16 | 2465 | 71 |
NM_001160233.1 | NP_001153705.1 | 15 - 16 | 2489 | 71 | |
NM_001160234.1 | NP_001153706.1 | 15 - 16 | 2298 | 71 | |
XM_017001360.1 | XP_016856849.1 | 16 - 17 | 5684 | 71 | |
XM_017001361.1 | XP_016856850.1 | 16 - 17 | 5026 | 71 | |
GenBank mRNA | AK293689.1 | - | 7 - 8 | 875 | 71 |
AK295095.1 | - | 15 - 16 | 2488 | 71 | |
AK296362.1 | - | 15 - 16 | 2298 | 71 | |
AK296384.1 | - | 15 - 16 | 2424 | 71 | |
AK308418.1 | - | 15 - 16 | 2230 | 71 | |
AK314777.1 | - | 15 - 16 | 2310 | 71 | |
BC001330.1 | - | 2 - 3 | 236 | 71 | |
BC003077.2 | - | 15 - 16 | 2378 | 71 | |
BC050359.1 | - | 15 - 16 | 2376 | 71 | |
J03007.1 | - | 10 - 11 | 1535 | 71 |