Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134262.2 | NP_599024.1 | 1 - 2 | 122 | 125 |
GenBank mRNA | AB209916.1 | - | 1 - 2 | 131 | 125 |
BC100987.2 | - | 1 - 2 | 72 | 125 | |
BC100988.1 | - | 1 - 2 | 72 | 125 | |
BC100989.1 | - | 1 - 2 | 72 | 125 | |
BC100990.2 | - | 1 - 2 | 72 | 125 |