Assay Design



Gene Transcripts

Gene Symbol
PSRC1
Entrez Gene ID
84722
Gene Name
proline and serine rich coiled-coil 1
Gene Aliases
DDA3, FP3214
Chromosome Location
Chr.1: 109279554 - 109283182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005290.3 NP_001005290.1 2 - 3 156 63
NM_001032291.2 NP_001027462.1 2 - 3 161 63
NM_032636.7 NP_116025.1 2 - 3 156 63
XM_005271283.2 XP_005271340.1 2 - 3 155 63
XM_011542306.2 XP_011540608.1 1 - 2 463 63
XM_017002560.1 XP_016858049.1 2 - 3 166 63
XM_017002561.1 XP_016858050.1 2 - 3 160 63
XM_017002562.1 XP_016858051.1 2 - 3 190 63
XM_017002563.1 XP_016858052.1 2 - 3 233 63
XM_017002564.1 XP_016858053.1 2 - 3 164 63
XM_017002565.1 XP_016858054.1 2 - 3 156 63
XM_017002566.1 XP_016858055.1 2 - 3 181 63
XM_017002567.1 XP_016858056.1 2 - 3 176 63
XM_017002568.1 XP_016858057.1 1 - 2 463 63
XM_017002569.1 XP_016858058.1 2 - 3 168 63
XM_017002570.1 XP_016858059.1 2 - 3 172 63
XM_017002571.1 XP_016858060.1 2 - 3 166 63
XM_017002572.1 XP_016858061.1 2 - 3 160 63
XM_017002573.1 XP_016858062.1 2 - 3 190 63
XM_017002574.1 XP_016858063.1 2 - 3 163 63
XM_017002575.1 XP_016858064.1 2 - 3 173 63
XM_017002576.1 XP_016858065.1 2 - 3 165 63
XM_017002577.1 XP_016858066.1 1 - 2 463 63
GenBank mRNA AF223000.1 - 2 - 3 135 63
AK126567.1 - 2 - 3 92 63
AK289593.1 - 2 - 3 98 63
BC056909.1 - 2 - 3 174 63
BF984061.1 - 2 - 3 137 63
BJ992594.1 - 2 - 3 161 63
BX649139.1 - 2 - 3 103 63

View Full Product Details