Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289006.1 | NP_001275935.1 | 5 - 6 | 1632 | 74 |
NM_006405.6 | NP_006396.2 | 5 - 6 | 1838 | 74 | |
GenBank mRNA | AK292110.1 | - | 5 - 6 | 1527 | 74 |
AK304642.1 | - | 5 - 6 | 1533 | 74 | |
BC010856.2 | - | 4 - 5 | 1447 | 74 | |
BX161390.1 | - | 5 - 6 | 1758 | 74 | |
U94831.1 | - | 4 - 5 | 1445 | 74 |