Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002553.3 | NP_002544.1 | 2 - 3 | 312 | 147 |
NM_181747.3 | NP_859531.1 | 2 - 3 | 312 | 147 | |
XM_011516273.2 | XP_011514575.1 | 2 - 3 | 306 | 147 | |
GenBank mRNA | AF047599.1 | - | 2 - 3 | 247 | 147 |
AF049127.1 | - | 2 - 3 | 243 | 147 | |
AF081459.1 | - | 2 - 3 | 222 | 147 | |
AK223361.1 | - | 2 - 3 | 312 | 147 | |
AK292285.1 | - | 2 - 3 | 288 | 147 | |
AK313596.1 | - | 2 - 3 | 269 | 147 | |
BC023652.2 | - | 2 - 3 | 250 | 147 | |
BF678849.1 | - | 2 - 3 | 145 | ||
U92538.1 | - | 2 - 3 | 257 | 147 |