Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002553.3 | NP_002544.1 | 4 - 5 | 585 | 121 |
NM_181747.3 | NP_859531.1 | 4 - 5 | 585 | 121 | |
XM_011516273.2 | XP_011514575.1 | 4 - 5 | 579 | 121 | |
GenBank mRNA | AF047599.1 | - | 4 - 5 | 520 | 121 |
AF049127.1 | - | 4 - 5 | 516 | 121 | |
AF081459.1 | - | 4 - 5 | 495 | 121 | |
AK223361.1 | - | 4 - 5 | 585 | 121 | |
AK302122.1 | - | 5 - 6 | 672 | 121 | |
AK313596.1 | - | 4 - 5 | 542 | 121 | |
BC023652.2 | - | 4 - 5 | 523 | 121 | |
U92538.1 | - | 4 - 5 | 530 | 121 |