Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002553.3 | NP_002544.1 | 6 - 7 | 824 | 144 |
NM_181747.3 | NP_859531.1 | 6 - 7 | 824 | 144 | |
GenBank mRNA | AF047599.1 | - | 6 - 7 | 759 | 144 |
AF049127.1 | - | 6 - 7 | 755 | 144 | |
AF081459.1 | - | 6 - 7 | 734 | 144 | |
AK223361.1 | - | 6 - 7 | 824 | 144 | |
AK292285.1 | - | 6 - 7 | 144 | ||
AK302122.1 | - | 7 - 8 | 911 | 144 | |
AK313596.1 | - | 6 - 7 | 781 | 144 | |
BC023652.2 | - | 6 - 7 | 762 | 144 | |
U92538.1 | - | 6 - 7 | 769 | 144 |