Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006187.3 | NP_006178.2 | 2 - 3 | 557 | 87 |
XM_005253889.3 | XP_005253946.1 | 2 - 3 | 478 | 87 | |
XM_017019363.1 | XP_016874852.1 | 2 - 3 | 478 | 87 | |
GenBank mRNA | AB044545.1 | - | 2 - 3 | 638 | 87 |
AF063613.1 | - | 2 - 3 | 493 | 87 | |
AK292949.1 | - | 2 - 3 | 527 | 87 | |
BC113746.1 | - | 2 - 3 | 532 | 87 |