Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006187.3 | NP_006178.2 | 8 - 9 | 1929 | 82 |
XM_005253889.3 | XP_005253946.1 | 8 - 9 | 1850 | 82 | |
XM_017019363.1 | XP_016874852.1 | 8 - 9 | 1850 | 82 | |
GenBank mRNA | AB044545.1 | - | 8 - 9 | 2010 | 82 |
AF063613.1 | - | 8 - 9 | 1865 | 82 | |
AK292949.1 | - | 8 - 9 | 1899 | 82 | |
BC113746.1 | - | 8 - 9 | 1904 | 82 | |
BM474470.1 | - | 3 - 4 | 627 | 82 |