Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040694.1 | NP_001035784.1 | 1 - 2 | 194 | 91 |
NM_020238.2 | NP_064623.2 | 1 - 2 | 194 | 91 | |
XM_006718533.2 | XP_006718596.1 | 1 - 2 | 156 | 91 | |
XM_011544995.2 | XP_011543297.1 | 1 - 2 | 156 | 91 | |
XM_011544996.2 | XP_011543298.1 | 1 - 2 | 156 | 91 | |
XM_011544997.2 | XP_011543299.1 | 1 - 2 | 116 | 91 | |
XM_011544998.2 | XP_011543300.1 | 1 - 2 | 156 | 91 | |
GenBank mRNA | AK056195.1 | - | 1 - 2 | 169 | 91 |
AK307633.1 | - | 1 - 2 | 91 | ||
AK310714.1 | - | 1 - 2 | 156 | 91 | |
BC032678.1 | - | 1 - 2 | 118 | 91 | |
BC098576.1 | - | 1 - 2 | 194 | 91 | |
CV569756.1 | - | 1 - 2 | 105 | 91 |