Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040694.1 | NP_001035784.1 | 4 - 5 | 1267 | 112 |
NM_020238.2 | NP_064623.2 | 4 - 5 | 1267 | 112 | |
XM_006718533.2 | XP_006718596.1 | 4 - 5 | 1229 | 112 | |
XM_011544995.2 | XP_011543297.1 | 4 - 5 | 1229 | 112 | |
XM_011544996.2 | XP_011543298.1 | 4 - 5 | 1229 | 112 | |
XM_011544997.2 | XP_011543299.1 | 4 - 5 | 1189 | 112 | |
XM_011544998.2 | XP_011543300.1 | 4 - 5 | 1229 | 112 | |
GenBank mRNA | AF282265.1 | - | 3 - 4 | 1065 | 112 |
AK056195.1 | - | 4 - 5 | 1242 | 112 | |
AK307633.1 | - | 4 - 5 | 1284 | 112 | |
AY714053.1 | - | 3 - 4 | 1065 | 112 | |
BC032678.1 | - | 4 - 5 | 1191 | 112 | |
BC098576.1 | - | 4 - 5 | 1267 | 112 |