Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005257.5 | NP_005248.2 | 3 - 4 | 1587 | 69 |
GenBank mRNA | AK223547.1 | - | 2 - 3 | 1516 | 69 |
D87811.1 | - | 2 - 3 | 1128 | 69 | |
U66075.1 | - | 2 - 3 | 1213 | 69 | |
X95701.1 | - | 3 - 4 | 1581 | 69 |