Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291916.1 | NP_001278845.1 | 1 - 2 | 175 | 83 |
NM_006938.3 | NP_008869.1 | 1 - 2 | 175 | 83 | |
GenBank mRNA | BC001721.1 | - | 1 - 2 | 136 | 83 |
BC072427.1 | - | 1 - 2 | 62 | 83 | |
BU565530.1 | - | 1 - 2 | 177 | 83 | |
CN354758.1 | - | 1 - 2 | 111 | 83 | |
HY043971.1 | - | 1 - 2 | 173 | 83 | |
J03798.1 | - | 1 - 2 | 164 | 83 |