Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289067.1 | NP_001275996.1 | 18 - 19 | 2266 | 91 |
NM_001289068.1 | NP_001275997.1 | 17 - 18 | 2083 | 91 | |
NM_001289069.1 | NP_001275998.1 | 17 - 18 | 2032 | 91 | |
NM_001289070.1 | NP_001275999.1 | 15 - 16 | 1834 | 91 | |
NM_001289071.1 | NP_001276000.1 | 18 - 19 | 2172 | 91 | |
NM_001289072.1 | NP_001276001.1 | 15 - 16 | 1738 | 91 | |
NM_001289073.1 | NP_001276002.1 | 16 - 17 | 2063 | 91 | |
NM_001289074.1 | NP_001276003.1 | 18 - 19 | 2047 | 91 | |
NM_001289075.1 | NP_001276004.1 | 16 - 17 | 1931 | 91 | |
NM_018063.4 | NP_060533.2 | 17 - 18 | 2128 | 91 | |
GenBank mRNA | AB074174.1 | - | 5 - 6 | 576 | 91 |
AB102716.1 | - | 18 - 19 | 2017 | 91 | |
AB102717.1 | - | 16 - 17 | 1908 | 91 | |
AB102718.1 | - | 17 - 18 | 1877 | 91 | |
AB102719.1 | - | 15 - 16 | 1583 | 91 | |
AB102720.1 | - | 16 - 17 | 1776 | 91 | |
AB102721.1 | - | 15 - 16 | 1679 | 91 | |
AB102722.1 | - | 18 - 19 | 1892 | 91 | |
AB102723.1 | - | 18 - 19 | 2111 | 91 | |
AB113249.1 | - | 17 - 18 | 1973 | 91 | |
AF155827.1 | - | 17 - 18 | 2078 | 91 | |
AK001201.1 | - | 3 - 4 | 286 | 91 | |
AK314485.1 | - | 17 - 18 | 2117 | 91 | |
AY007108.1 | - | 5 - 6 | 585 | 91 | |
BC015477.1 | - | 4 - 5 | 411 | 91 | |
BC029381.1 | - | 5 - 6 | 516 | 91 | |
BC030963.1 | - | 6 - 7 | 731 | 91 | |
BC031004.1 | - | 6 - 7 | 731 | 91 | |
BX538033.1 | - | 17 - 18 | 2067 | 91 |