Assay Design



Gene Transcripts

Gene Symbol
HELLS
Entrez Gene ID
3070
Gene Name
helicase, lymphoid-specific
Gene Aliases
ICF4, LSH, Nbla10143, PASG, SMARCA6
Chromosome Location
Chr.10: 94545767 - 94613905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289067.1 NP_001275996.1 21 - 22 2630 122
NM_001289068.1 NP_001275997.1 20 - 21 2447 122
NM_001289069.1 NP_001275998.1 20 - 21 2396 122
NM_001289070.1 NP_001275999.1 18 - 19 2198 122
NM_001289071.1 NP_001276000.1 21 - 22 2536 122
NM_001289072.1 NP_001276001.1 18 - 19 2102 122
NM_001289073.1 NP_001276002.1 19 - 20 2427 122
NM_001289074.1 NP_001276003.1 21 - 22 2411 122
NM_001289075.1 NP_001276004.1 19 - 20 2295 122
NM_018063.4 NP_060533.2 20 - 21 2492 122
GenBank mRNA AB074174.1 - 8 - 9 940 122
AB102716.1 - 21 - 22 2381 122
AB102717.1 - 19 - 20 2272 122
AB102718.1 - 20 - 21 2241 122
AB102719.1 - 18 - 19 1947 122
AB102720.1 - 19 - 20 2140 122
AB102721.1 - 18 - 19 2043 122
AB102722.1 - 21 - 22 2256 122
AB102723.1 - 21 - 22 2475 122
AB113248.1 - 20 - 21 2250 122
AF155827.1 - 20 - 21 2442 122
AK001201.1 - 6 - 7 650 122
AK314485.1 - 20 - 21 2481 122
AY007108.1 - 8 - 9 949 122
BC015477.1 - 7 - 8 775 122
BC029381.1 - 8 - 9 880 122
BC030963.1 - 9 - 10 1095 122
BC031004.1 - 9 - 10 1095 122
BX538033.1 - 20 - 21 2431 122

View Full Product Details