Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282948.1 | NP_001269877.1 | 15 - 16 | 124 | |
NM_001318153.1 | NP_001305082.1 | 15 - 16 | 124 | ||
NM_001318154.1 | NP_001305083.1 | 16 - 17 | 124 | ||
NM_001322252.1 | NP_001309181.1 | 15 - 16 | 124 | ||
NM_001322254.1 | NP_001309183.1 | 14 - 15 | 124 | ||
NM_001322258.1 | NP_001309187.1 | 15 - 16 | 124 | ||
NM_032776.2 | NP_116165.1 | 16 - 17 | 124 | ||
NR_134512.1 | 12 - 13 | 124 | |||
XM_011539503.1 | XP_011537805.1 | 15 - 16 | 124 | ||
XM_017015897.1 | XP_016871386.1 | 15 - 16 | 124 | ||
XM_017015898.1 | XP_016871387.1 | 16 - 17 | 124 | ||
XM_017015899.1 | XP_016871388.1 | 16 - 17 | 124 | ||
XM_017015900.1 | XP_016871389.1 | 16 - 17 | 124 | ||
XM_017015901.1 | XP_016871390.1 | 15 - 16 | 124 | ||
XM_017015902.1 | XP_016871391.1 | 15 - 16 | 124 | ||
XM_017015903.1 | XP_016871392.1 | 14 - 15 | 124 | ||
GenBank mRNA | AB037801.1 | - | 7 - 8 | 124 | |
AK024991.1 | - | 5 - 6 | 1716 | 123 | |
AL831917.1 | - | 9 - 10 | 124 | ||
BC143722.1 | - | 15 - 16 | 124 | ||
BX537954.1 | - | 12 - 13 | 124 | ||
EF068222.1 | - | 18 - 19 | 124 |