Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021942.5 | NP_068761.4 | 14 - 15 | 1620 | 99 |
NM_199053.2 | NP_951008.1 | 14 - 15 | 1620 | 99 | |
XM_017008537.1 | XP_016864026.1 | 14 - 15 | 1620 | 99 | |
XM_017008538.1 | XP_016864027.1 | 14 - 15 | 1620 | 99 | |
XM_017008539.1 | XP_016864028.1 | 12 - 13 | 1271 | 99 | |
GenBank mRNA | AK091146.1 | - | 9 - 10 | 844 | 99 |
AK096345.1 | - | 14 - 15 | 1594 | 99 | |
AK315057.1 | - | 9 - 10 | 826 | 99 | |
AL136752.1 | - | 6 - 7 | 529 | 99 | |
AL833451.1 | - | 13 - 14 | 1428 | 99 | |
AL833571.1 | - | 14 - 15 | 1599 | 99 | |
BC051724.1 | - | 14 - 15 | 1564 | 99 | |
BX538044.1 | - | 14 - 15 | 1574 | 99 | |
BX647127.1 | - | 14 - 15 | 1591 | 99 | |
BX648499.1 | - | 14 - 15 | 1595 | 99 | |
CR533553.1 | - | 6 - 7 | 464 | 99 |