Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021942.5 | NP_068761.4 | 24 - 25 | 2902 | 77 |
NM_199053.2 | NP_951008.1 | 24 - 25 | 2902 | 77 | |
XM_017008537.1 | XP_016864026.1 | 24 - 25 | 2902 | 77 | |
XM_017008538.1 | XP_016864027.1 | 24 - 25 | 2902 | 77 | |
XM_017008539.1 | XP_016864028.1 | 22 - 23 | 2553 | 77 | |
GenBank mRNA | AK022778.1 | - | 3 - 4 | 306 | 77 |
AK023390.1 | - | 5 - 6 | 591 | 77 | |
AK026992.1 | - | 10 - 11 | 1294 | 77 | |
AK091146.1 | - | 19 - 20 | 2126 | 77 | |
AK096345.1 | - | 24 - 25 | 2876 | 77 | |
AK315057.1 | - | 19 - 20 | 2108 | 77 | |
AL136752.1 | - | 16 - 17 | 1811 | 77 | |
AL833451.1 | - | 23 - 24 | 2708 | 77 | |
AL833571.1 | - | 24 - 25 | 2881 | 77 | |
BC051724.1 | - | 24 - 25 | 2846 | 77 | |
BC139745.1 | - | 12 - 13 | 1282 | 77 | |
BX538044.1 | - | 24 - 25 | 2856 | 77 | |
BX647127.1 | - | 24 - 25 | 2873 | 77 | |
BX648499.1 | - | 24 - 25 | 2877 | 77 | |
CR533553.1 | - | 16 - 17 | 1746 | 77 |