Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021942.5 | NP_068761.4 | 8 - 9 | 1034 | 123 |
NM_199053.2 | NP_951008.1 | 8 - 9 | 1034 | 123 | |
XM_017008537.1 | XP_016864026.1 | 8 - 9 | 1034 | 123 | |
XM_017008538.1 | XP_016864027.1 | 8 - 9 | 1034 | 123 | |
XM_017008539.1 | XP_016864028.1 | 7 - 8 | 779 | 123 | |
GenBank mRNA | AK091146.1 | - | 3 - 4 | 258 | 123 |
AK096345.1 | - | 8 - 9 | 1008 | 123 | |
AK315057.1 | - | 3 - 4 | 240 | 123 | |
AL833451.1 | - | 7 - 8 | 842 | 123 | |
AL833571.1 | - | 8 - 9 | 1013 | 123 | |
BC051724.1 | - | 8 - 9 | 978 | 123 | |
BX538044.1 | - | 8 - 9 | 988 | 123 | |
BX647127.1 | - | 8 - 9 | 1005 | 123 | |
BX648499.1 | - | 8 - 9 | 1009 | 123 |