Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020453.3 | NP_065186.3 | 18 - 19 | 3533 | 76 |
XM_005248119.4 | XP_005248176.1 | 18 - 19 | 3619 | 76 | |
XM_005248120.4 | XP_005248177.1 | 18 - 19 | 3651 | 76 | |
XM_011513722.2 | XP_011512024.1 | 17 - 18 | 3469 | 76 | |
XM_017008472.1 | XP_016863961.1 | 19 - 20 | 3753 | 76 | |
GenBank mRNA | AB040920.1 | - | 7 - 8 | 1043 | 76 |
AJ441078.1 | - | 18 - 19 | 3533 | 76 | |
AK027598.1 | - | 11 - 12 | 1724 | 76 | |
AK074930.1 | - | 1 - 2 | 51 | 76 | |
BC131535.1 | - | 18 - 19 | 3511 | 76 |