Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020453.3 | NP_065186.3 | 22 - 23 | 4101 | 96 |
XM_005248119.4 | XP_005248176.1 | 22 - 23 | 4187 | 96 | |
XM_005248120.4 | XP_005248177.1 | 21 - 22 | 4138 | 96 | |
XM_011513722.2 | XP_011512024.1 | 21 - 22 | 4037 | 96 | |
XM_017008472.1 | XP_016863961.1 | 23 - 24 | 4321 | 96 | |
GenBank mRNA | AB040920.1 | - | 11 - 12 | 1611 | 96 |
AJ441078.1 | - | 22 - 23 | 4101 | 96 | |
AK027598.1 | - | 15 - 16 | 2292 | 96 | |
AK074930.1 | - | 5 - 6 | 619 | 96 | |
BC131535.1 | - | 22 - 23 | 4079 | 96 |