Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002106.3 | NP_002097.1 | 3 - 4 | 379 | 62 |
GenBank mRNA | AK315413.1 | - | 3 - 4 | 297 | 62 |
AY597416.1 | - | 3 - 4 | 316 | 62 | |
BC018002.1 | - | 3 - 4 | 299 | 62 | |
BC020936.1 | - | 3 - 4 | 299 | 62 | |
BC103743.1 | - | 3 - 4 | 414 | 62 | |
CD685635.1 | - | 3 - 4 | 412 | 62 | |
CR457415.1 | - | 2 - 3 | 194 | 62 | |
M37583.1 | - | 3 - 4 | 300 | 62 | |
X52317.1 | - | 3 - 4 | 300 | 62 |