Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135648.2 | NP_001129120.1 | 13 - 14 | 2513 | 73 |
NM_001291981.1 | NP_001278910.1 | 13 - 14 | 2513 | 73 | |
NM_001291982.1 | NP_001278911.1 | 13 - 14 | 2513 | 73 | |
NM_001291983.1 | NP_001278912.1 | 12 - 13 | 2241 | 73 | |
NM_002844.3 | NP_002835.2 | 13 - 14 | 2513 | 73 | |
XM_011536015.2 | XP_011534317.1 | 13 - 14 | 2564 | 73 | |
XM_011536016.2 | XP_011534318.1 | 13 - 14 | 2206 | 73 | |
XM_011536017.2 | XP_011534319.1 | 14 - 15 | 2804 | 73 | |
XM_011536018.2 | XP_011534320.1 | 14 - 15 | 2477 | 73 | |
XM_017011145.1 | XP_016866634.1 | 14 - 15 | 2528 | 73 | |
XM_017011146.1 | XP_016866635.1 | 14 - 15 | 2477 | 73 | |
GenBank mRNA | AB209670.1 | - | 11 - 12 | 1754 | 73 |
AF533875.1 | - | 13 - 14 | 2278 | 73 | |
AK295028.1 | - | 13 - 14 | 2438 | 73 | |
BC140775.1 | - | 13 - 14 | 2315 | 73 | |
BC144512.1 | - | 13 - 14 | 2315 | 73 | |
BC144513.1 | - | 13 - 14 | 2315 | 73 | |
CR749277.1 | - | 13 - 14 | 2454 | 73 | |
L77886.1 | - | 13 - 14 | 2212 | 73 |