Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135648.2 | NP_001129120.1 | 30 - 31 | 4606 | 70 |
NM_001291981.1 | NP_001278910.1 | 32 - 33 | 4654 | 70 | |
NM_001291984.1 | NP_001278913.1 | 29 - 30 | 4585 | 70 | |
NM_002844.3 | NP_002835.2 | 29 - 30 | 4588 | 70 | |
XM_006715537.2 | XP_006715600.1 | 30 - 31 | 4654 | 70 | |
XM_011536014.2 | XP_011534316.1 | 32 - 33 | 4702 | 70 | |
XM_011536015.2 | XP_011534317.1 | 31 - 32 | 4693 | 70 | |
XM_011536016.2 | XP_011534318.1 | 31 - 32 | 4329 | 70 | |
XM_011536017.2 | XP_011534319.1 | 33 - 34 | 4945 | 70 | |
XM_011536018.2 | XP_011534320.1 | 33 - 34 | 4618 | 70 | |
XM_017011145.1 | XP_016866634.1 | 31 - 32 | 4621 | 70 | |
XM_017011146.1 | XP_016866635.1 | 31 - 32 | 4570 | 70 | |
XM_017011147.1 | XP_016866636.1 | 31 - 32 | 4548 | 70 | |
XM_017011148.1 | XP_016866637.1 | 30 - 31 | 4527 | 70 | |
XM_017011149.1 | XP_016866638.1 | 20 - 21 | 2206 | 70 | |
XM_017011150.1 | XP_016866639.1 | 19 - 20 | 2193 | 70 | |
GenBank mRNA | AF533875.1 | - | 29 - 30 | 4353 | 70 |
BC140775.1 | - | 30 - 31 | 4408 | 70 | |
BC144512.1 | - | 30 - 31 | 4408 | 70 | |
BC144513.1 | - | 32 - 33 | 4456 | 70 | |
CR749277.1 | - | 29 - 30 | 4529 | 70 | |
L77886.1 | - | 29 - 30 | 4287 | 70 | |
Z70660.1 | - | 29 - 30 | 4269 | 70 |