Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002553.3 | NP_002544.1 | 10 - 11 | 1131 | 109 |
GenBank mRNA | AF047599.1 | - | 10 - 11 | 1066 | 109 |
AF049127.1 | - | 10 - 11 | 1062 | 109 | |
AK223361.1 | - | 10 - 11 | 1131 | 109 | |
AK292285.1 | - | 10 - 11 | 1107 | 109 | |
AK302122.1 | - | 11 - 12 | 1218 | 109 | |
AK313596.1 | - | 10 - 11 | 1088 | 109 | |
BC023652.2 | - | 10 - 11 | 1069 | 109 | |
U92538.1 | - | 10 - 11 | 1076 | 109 |