Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005576.3 | NP_005567.2 | 3 - 4 | 1676 | 92 |
XM_017022179.1 | XP_016877668.1 | 3 - 4 | 692 | 92 | |
GenBank mRNA | AK303982.1 | - | 4 - 5 | 1280 | 92 |
AK314222.1 | - | 3 - 4 | 1674 | 92 | |
BC015090.2 | - | 3 - 4 | 1655 | 92 | |
BC068542.1 | - | 3 - 4 | 1676 | 92 | |
L21186.1 | - | 3 - 4 | 1655 | 92 |