Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278469.1 | NP_001265398.1 | 31 - 32 | 4968 | 151 |
NM_001278470.1 | NP_001265399.1 | 30 - 31 | 4950 | 151 | |
NM_001278471.1 | NP_001265400.1 | 30 - 31 | 4881 | 151 | |
NM_001278472.1 | NP_001265401.1 | 32 - 33 | 5046 | 151 | |
NM_015026.2 | NP_055841.2 | 31 - 32 | 4968 | 151 | |
XM_017019041.1 | XP_016874530.1 | 32 - 33 | 4971 | 151 | |
XM_017019042.1 | XP_016874531.1 | 31 - 32 | 4953 | 151 | |
XM_017019043.1 | XP_016874532.1 | 33 - 34 | 5045 | 151 | |
XM_017019044.1 | XP_016874533.1 | 33 - 34 | 5047 | 151 | |
XM_017019045.1 | XP_016874534.1 | 34 - 35 | 4715 | 151 | |
XM_017019046.1 | XP_016874535.1 | 28 - 29 | 4140 | 151 | |
GenBank mRNA | AB017814.1 | - | 31 - 32 | 4693 | 151 |
AB028963.3 | - | 32 - 33 | 4939 | 151 | |
AK022671.1 | - | 5 - 6 | 1145 | 151 | |
AK092646.1 | - | 5 - 6 | 1732 | 151 | |
AL833066.2 | - | 31 - 32 | 4963 | 151 | |
AL834320.1 | - | 25 - 26 | 3926 | 151 | |
BC136621.1 | - | 30 - 31 | 4919 | 151 | |
BC144309.1 | - | 30 - 31 | 4850 | 151 | |
BC151241.1 | - | 32 - 33 | 4962 | 151 | |
BX537415.1 | - | 31 - 32 | 4966 | 151 |