Assay Design



Gene Transcripts

Gene Symbol
TECTB
Entrez Gene ID
6975
Gene Name
tectorin beta
Gene Aliases
-
Chromosome Location
Chr.10: 112283656 - 112305035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058222.2 NP_478129.1 8 - 9 989 127
GenBank mRNA AF312827.1 - 8 - 9 910 127
BC113497.1 - 8 - 9 989 127
BC113499.1 - 8 - 9 989 127

View Full Product Details