Assay Design



Gene Transcripts

Gene Symbol
KLHL8
Entrez Gene ID
57563
Gene Name
kelch like family member 8
Gene Aliases
-
Chromosome Location
Chr.4: 87160103 - 87220608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292003.1 NP_001278932.1 6 - 7 111
NM_001292006.1 NP_001278935.1 6 - 7 111
NM_001292007.1 NP_001278936.1 5 - 6 111
NM_020803.4 NP_065854.3 6 - 7 111
NR_120447.1 4 - 5 111
GenBank mRNA AB037799.2 - 5 - 6 111
AK128827.1 - 6 - 7 111
BC041384.2 - 6 - 7 1507 111
BC041901.1 - 4 - 5 111
BX640727.1 - 6 - 7 111
BX640744.1 - 6 - 7 111
BX647454.1 - 6 - 7 111
BX648367.1 - 4 - 5 111
CR749265.1 - 5 - 6 111

View Full Product Details