Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005038.2 | NP_005029.1 | 6 - 7 | 862 | 185 |
GenBank mRNA | AK313929.1 | - | 6 - 7 | 863 | 185 |
BC030707.2 | - | 6 - 7 | 858 | 185 | |
CR542081.1 | - | 6 - 7 | 750 | 185 | |
L11667.1 | - | 6 - 7 | 849 | 185 |