Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243084.1 | NP_001230013.1 | 3 - 4 | 672 | 150 |
NM_001530.3 | NP_001521.1 | 3 - 4 | 775 | 150 | |
NM_181054.2 | NP_851397.1 | 3 - 4 | 775 | 150 | |
GenBank mRNA | AB073325.1 | - | 3 - 4 | 399 | 150 |
AB500182.1 | - | 3 - 4 | 396 | 150 | |
AF207601.1 | - | 3 - 4 | 381 | 150 | |
AF207602.1 | - | 3 - 4 | 378 | 150 | |
AF304431.1 | - | 3 - 4 | 371 | 150 | |
AK299551.1 | - | 4 - 5 | 477 | 150 | |
AK304436.1 | - | 2 - 3 | 472 | 150 | |
BC012527.2 | - | 3 - 4 | 625 | 150 | |
BT009776.1 | - | 3 - 4 | 371 | 150 | |
BX648795.1 | - | 3 - 4 | 639 | 150 | |
FJ790247.1 | - | 3 - 4 | 673 | 150 | |
U22431.1 | - | 3 - 4 | 399 | 150 | |
U29165.1 | - | 3 - 4 | 635 | 150 |