Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020343.3 | NP_065076.2 | 19 - 20 | 2670 | 68 |
XM_005260768.2 | XP_005260825.1 | 19 - 20 | 2685 | 68 | |
XM_006723598.1 | XP_006723661.1 | 20 - 21 | 2826 | 68 | |
XM_006723599.1 | XP_006723662.1 | 20 - 21 | 2826 | 68 | |
XM_011529309.1 | XP_011527611.1 | 20 - 21 | 2826 | 68 | |
XM_011529310.2 | XP_011527612.1 | 18 - 19 | 2447 | 68 | |
XM_011529311.2 | XP_011527613.2 | 6 - 7 | 2786 | 68 | |
XM_017027972.1 | XP_016883461.1 | 20 - 21 | 2826 | 68 | |
XM_017027973.1 | XP_016883462.1 | 17 - 18 | 2289 | 68 | |
XM_017027974.1 | XP_016883463.1 | 17 - 18 | 2284 | 68 | |
XM_017027975.1 | XP_016883464.1 | 19 - 20 | 2618 | 68 | |
XM_017027976.1 | XP_016883465.1 | 17 - 18 | 2471 | 68 | |
XM_017027977.1 | XP_016883466.1 | 20 - 21 | 2826 | 68 | |
GenBank mRNA | AK002211.1 | - | 11 - 12 | 1540 | 68 |
BC171842.1 | - | 17 - 18 | 2471 | 68 |