Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020343.3 | NP_065076.2 | 25 - 26 | 3552 | 96 |
XM_005260768.2 | XP_005260825.1 | 25 - 26 | 3567 | 96 | |
XM_006723598.1 | XP_006723661.1 | 26 - 27 | 3708 | 96 | |
XM_006723599.1 | XP_006723662.1 | 26 - 27 | 3708 | 96 | |
XM_011529309.1 | XP_011527611.1 | 26 - 27 | 3708 | 96 | |
XM_011529310.2 | XP_011527612.1 | 24 - 25 | 3329 | 96 | |
XM_011529311.2 | XP_011527613.2 | 12 - 13 | 3668 | 96 | |
XM_017027972.1 | XP_016883461.1 | 26 - 27 | 3708 | 96 | |
XM_017027973.1 | XP_016883462.1 | 23 - 24 | 3171 | 96 | |
XM_017027974.1 | XP_016883463.1 | 23 - 24 | 3166 | 96 | |
XM_017027975.1 | XP_016883464.1 | 25 - 26 | 3500 | 96 | |
XM_017027976.1 | XP_016883465.1 | 23 - 24 | 3353 | 96 | |
GenBank mRNA | AK002211.1 | - | 17 - 18 | 2422 | 95 |
BC171842.1 | - | 23 - 24 | 3353 | 96 | |
DQ310704.1 | - | 25 - 26 | 3409 | 96 |