Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020343.3 | NP_065076.2 | 35 - 36 | 5354 | 83 |
XM_005260768.2 | XP_005260825.1 | 35 - 36 | 5369 | 83 | |
XM_006723598.1 | XP_006723661.1 | 36 - 37 | 5510 | 83 | |
XM_006723599.1 | XP_006723662.1 | 36 - 37 | 5510 | 83 | |
XM_011529309.1 | XP_011527611.1 | 36 - 37 | 5510 | 83 | |
XM_011529310.2 | XP_011527612.1 | 34 - 35 | 5131 | 83 | |
XM_011529311.2 | XP_011527613.2 | 22 - 23 | 5470 | 83 | |
XM_017027972.1 | XP_016883461.1 | 36 - 37 | 5510 | 83 | |
XM_017027973.1 | XP_016883462.1 | 33 - 34 | 4973 | 83 | |
XM_017027974.1 | XP_016883463.1 | 33 - 34 | 4968 | 83 | |
XM_017027975.1 | XP_016883464.1 | 35 - 36 | 5302 | 83 | |
XM_017027976.1 | XP_016883465.1 | 33 - 34 | 5155 | 83 | |
GenBank mRNA | AK002211.1 | - | 27 - 28 | 4223 | 83 |
AK022881.1 | - | 4 - 5 | 443 | 83 | |
AK127228.1 | - | 8 - 9 | 3525 | 83 | |
BC013749.1 | - | 4 - 5 | 502 | 83 | |
BC171842.1 | - | 33 - 34 | 5155 | 83 | |
DQ310704.1 | - | 35 - 36 | 5211 | 83 |