Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003384.2 | NP_003375.1 | 1 - 2 | 100 | 119 |
XM_006720247.3 | XP_006720310.1 | 1 - 2 | 138 | 119 | |
XM_017021624.1 | XP_016877113.1 | 1 - 2 | 138 | 119 | |
XM_017021626.1 | XP_016877115.1 | 1 - 2 | 138 | 119 | |
GenBank mRNA | AB000449.1 | - | 1 - 2 | 69 | 119 |
AK290110.1 | - | 1 - 2 | 88 | 119 | |
BC103761.1 | - | 1 - 2 | 78 | 119 | |
BC112075.1 | - | 1 - 2 | 68 | 119 | |
BC113510.1 | - | 1 - 2 | 68 | 119 | |
BU535261.1 | - | 1 - 2 | 119 |