Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114.4 | NP_001105.1 | 16 - 17 | 2336 | 57 |
NM_001286057.1 | NP_001272986.1 | 17 - 18 | 2384 | 57 | |
XM_011522835.2 | XP_011521137.1 | 17 - 18 | 2437 | 57 | |
XM_011522836.2 | XP_011521138.1 | 17 - 18 | 2453 | 57 | |
XM_011522837.2 | XP_011521139.1 | 17 - 18 | 2382 | 57 | |
XM_011522838.2 | XP_011521140.1 | 17 - 18 | 2586 | 57 | |
XM_011522839.2 | XP_011521141.1 | 17 - 18 | 2413 | 57 | |
XM_011522840.2 | XP_011521142.1 | 17 - 18 | 2420 | 57 | |
XM_011522842.2 | XP_011521144.1 | 17 - 18 | 2483 | 57 | |
XM_017022895.1 | XP_016878384.1 | 17 - 18 | 2453 | 57 | |
XM_017022896.1 | XP_016878385.1 | 14 - 15 | 1599 | 57 | |
XM_017022897.1 | XP_016878386.1 | 14 - 15 | 1618 | 57 | |
XM_017022898.1 | XP_016878387.1 | 17 - 18 | 2453 | 57 | |
XM_017022899.1 | XP_016878388.1 | 17 - 18 | 2453 | 57 | |
GenBank mRNA | AK093706.1 | - | 18 - 19 | 2334 | 57 |
BC039891.1 | - | 14 - 15 | 1972 | 57 | |
BC126271.1 | - | 16 - 17 | 2148 | 57 | |
D25538.1 | - | 16 - 17 | 2333 | 57 |