Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020884.4 | NP_065935.3 | 20 - 21 | 2275 | 56 |
XM_006723840.3 | XP_006723903.1 | 18 - 19 | 1916 | 56 | |
XM_011528941.2 | XP_011527243.1 | 18 - 19 | 1940 | 56 | |
XM_011528947.2 | XP_011527249.1 | 12 - 13 | 2313 | 56 | |
XM_011528948.2 | XP_011527250.1 | 12 - 13 | 1472 | 56 | |
XM_017027986.1 | XP_016883475.1 | 19 - 20 | 1976 | 56 | |
XM_017027987.1 | XP_016883476.1 | 14 - 15 | 1491 | 56 | |
GenBank mRNA | AB040945.2 | - | 18 - 19 | 1984 | 56 |
BC151242.1 | - | 18 - 19 | 1996 | 56 |